Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB15546

Sigma-Aldrich

Anti-MAML1 Antibody

serum, Chemicon®

Synonym(s):

MAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAML1(9794)

General description

Members of the evolutionarily conserved Mastermind (MAM) protein family, including the three related mammalian Mastermind-like (MAML) proteins MAML1–3, function as crucial coactivators of Notch-mediated transcriptional activation. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.

Specificity

Cat. # AB15546 recognizes MAML1 protein.
Reactivity with other species has not been tested.

Immunogen

Synthetic Linear Peptide.

Application

MAML1 can be detected at 1/1000 dilution in 10 µg of HEK293 cell Lysate.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-MAML1 Antibody is validated for use in WB for the detection of MAML1.

Quality

Routinely evaluated by immunoblot.

Target description

108 kDa

Physical form

Serum with 0.05% NaN3

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
Tested by Western Blot on HEK293 cell lysate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service