Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-552

Sigma-Aldrich

TK Substrate 1

TK Substrate 1 primarily used in Kinase Assays, liquid

Sign Into View Organizational & Contract Pricing

Select a Size

297 μL
$334.00

$334.00


Estimated to ship onApril 24, 2025



Select a Size

Change View
297 μL
$334.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.42

$334.00


Estimated to ship onApril 24, 2025


Product Name

TK Substrate 1, TK Substrate 1 primarily used in Kinase Assays.

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

dry ice

Biochem/physiol Actions

Protein Target: TYK2

Quality

routinely used as a tyrosine kinase substrate with KinEASE fluorescence polarization assays

Storage and Stability

stable 6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service