Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191183

Sigma-Aldrich

3,5,5-Trimethyl-2-cyclohexen-1-ol

96%

Synonym(s):

Isophorol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H6OH
CAS Number:
Molecular Weight:
140.22
Beilstein/REAXYS Number:
1929064
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.472 (lit.)

bp

79.5-81.5 °C/8 mmHg (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CC1=CC(O)CC(C)(C)C1

InChI

1S/C9H16O/c1-7-4-8(10)6-9(2,3)5-7/h4,8,10H,5-6H2,1-3H3

InChI key

LDRWAWZXDDBHTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5,5-Trimethyl-2-cyclohexen-1-ol undergoes oxidation in the presence of palladium on activated charcoal under an ethylene atmosphere to yield the corresponding carbonyl compound.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

176.0 °F - closed cup

flash_point_c

80 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmentally benign oxidation using a palladium catalyst system.
Hayashi M, et al.
Green Chemistry, 2(6), 257-260 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service