Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900528

Sigma-Aldrich

2,2-Bis(hydroxymethyl)butyric acid

Vetec, reagent grade, 98%

Synonym(s):

2,2-Bis(hydroxymethyl)butanoic acid, 2,2-Bis(methylol)butanoic acid, 2,2-Dimethylolbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH2OH)2CO2H
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

109-112 °C (lit.)

SMILES string

CCC(CO)(CO)C(O)=O

InChI

1S/C6H12O4/c1-2-6(3-7,4-8)5(9)10/h7-8H,2-4H2,1H3,(H,9,10)

InChI key

JVYDLYGCSIHCMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service