Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46871-U

Supelco

Acrolein/Acrylonitrile Mix

10,000 μg/mL in deionized water, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

agency

EPA 8260

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

10,000 μg/mL in deionized water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Acrolein/Acrylonitrile Mix (Method: EPA 8260) is utilized for identifying and quantifying volatile organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8260 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acrolein
    Acrylonitrile

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service