Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20
EC Number:
201-997-7
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
140443
MDL number:
grade
purum
assay
≥97.0% (HPLC)
impurities
≤0.5% xanthene
bp
349-350 °C/730 mmHg (lit.)
mp
172-174 °C (lit.), 172-177 °C
SMILES string
O=C1c2ccccc2Oc3ccccc13
InChI
1S/C13H8O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H
InChI key
JNELGWHKGNBSMD-UHFFFAOYSA-N
Gene Information
mouse ... Prkch(18755)
Still not finding the right product?
Explore all of our products under Xanthone
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.