Skip to Content
MilliporeSigma

94837

(±)-Verapamil hydrochloride

≥99.0% (TLC)

Synonym(s):

5-[N-(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2N(CH3)(CH2)3C[C6H3(OCH3)2][CH(CH3)2]CN · HCl
CAS Number:
Molecular Weight:
491.06
EC Number:
205-800-5
UNSPSC Code:
12352203
PubChem Substance ID:
Beilstein/REAXYS Number:
3647093
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (TLC)

mp

142 °C (dec.) (lit.)

solubility

methanol: 50 mg/mL, clear, colorless

SMILES string

Cl.COc1ccc(CCN(C)CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC

InChI

1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H

InChI key

DOQPXTMNIUCOSY-UHFFFAOYSA-N

Biochem/physiol Actions

α1-adrenoceptor antagonist; L-type calcium channel blocker.
α1-adrenoceptor antagonist; L-type calcium channel blocker. Blocks L-type Ca2+ channels in smooth and cardiac muscle, induces apoptosis of human primary and metastatic colon adenocarcinoma cells in vitro. Drug resistance reversal agent acting on Pgp, e.g. decrease renal tubule elimination of digoxin. Increases basal ATPase activity of Pgp. Substrate of Cyp3A4 and CYP2C6.


Still not finding the right product?

Explore all of our products under (±)-Verapamil hydrochloride




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library