Skip to Content

92780

Glyceryl trimyristate

purum, ≥97.0% (GC)

Synonym(s):

1,2,3-Tritetradecanoylglycerol, Glycerol trimyristate, Trimyristin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3(CH2)12COOCH2]2CHOCO(CH2)12CH3
CAS Number:
Molecular Weight:
723.16
EC Number:
209-099-7
UNSPSC Code:
12352211
MDL number:
Beilstein/REAXYS Number:
1718242
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (GC)

mp

56-57 °C (lit.)

SMILES string

CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

InChI

1S/C45H86O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-43(46)49-40-42(51-45(48)39-36-33-30-27-24-21-18-15-12-9-6-3)41-50-44(47)38-35-32-29-26-23-20-17-14-11-8-5-2/h42H,4-41H2,1-3H3

InChI key

DUXYWXYOBMKGIN-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library