Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
EC Number:
202-122-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1864778
MDL number:
grade
purum
assay
≥99.0% (GC)
bp
379 °C (lit.)
mp
84-88 °C (lit.)
SMILES string
c1ccc(cc1)-c2cccc(c2)-c3ccccc3
InChI
1S/C18H14/c1-3-8-15(9-4-1)17-12-7-13-18(14-17)16-10-5-2-6-11-16/h1-14H
InChI key
YJTKZCDBKVTVBY-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

