Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
EC Number:
205-754-6
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1209898
grade
purum
assay
≥97.0% (GC)
refractive index
n20/D 1.552
bp
113-115 °C/5 mmHg (lit.)
density
1.131 g/mL at 25 °C (lit.)
SMILES string
COc1cccc(O)c1
InChI
1S/C7H8O2/c1-9-7-4-2-3-6(8)5-7/h2-5,8H,1H3
InChI key
ASHGTJPOSUFTGB-UHFFFAOYSA-N
Disclaimer
may discolor yellow to red on storage
Still not finding the right product?
Explore all of our products under 3-Methoxyphenol
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.