Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
EC Number:
200-639-7
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
152784
assay
≥99.0% (HPLC)
mp
160-162 °C
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
O=C1Oc2cc3occc3cc2C=C1
InChI
1S/C11H6O3/c12-11-2-1-7-5-8-3-4-13-9(8)6-10(7)14-11/h1-6H
InChI key
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Application
Photochemical reagent for the investigation of nucleic acid structure and function.
Still not finding the right product?
Explore all of our products under Psoralen
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
