Skip to Content
MilliporeSigma

78540

Methyl phenylacetate

purum, ≥99.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2CO2CH3
CAS Number:
Molecular Weight:
150.17
EC Number:
202-940-9
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
878795
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥99.0% (GC)

refractive index

n20/D 1.503 (lit.), n20/D 1.507

bp

218 °C (lit.)

density

1.066 g/mL at 20 °C (lit.)

SMILES string

COC(=O)Cc1ccccc1

InChI

1S/C9H10O2/c1-11-9(10)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

CRZQGDNQQAALAY-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Methyl phenylacetate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library