Skip to Content
MilliporeSigma

75770

Oxamide

purum, ≥98.0% (N)

Synonym(s):

Oxalic acid diamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2COCONH2
CAS Number:
Molecular Weight:
88.07
EC Number:
207-442-5
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1743262
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (N)

mp

>300 °C (lit.)

SMILES string

NC(=O)C(N)=O

InChI

1S/C2H4N2O2/c3-1(5)2(4)6/h(H2,3,5)(H2,4,6)

InChI key

YIKSCQDJHCMVMK-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library