Skip to Content
MilliporeSigma

75420

Orcinol monohydrate

purum, ≥96.0% (GC)

Synonym(s):

3,5-Dihydroxytoluene, 5-Methylresorcinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3C6H3-1,3-(OH)2 · H2O
CAS Number:
Molecular Weight:
142.15
EC Number:
207-984-2
UNSPSC Code:
12171500
PubChem Substance ID:
Beilstein/REAXYS Number:
1071903
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥96.0% (GC)

bp

290 °C (lit.)

mp

56-58 °C (lit.)

SMILES string

O.Cc1cc(O)cc(O)c1

InChI

1S/C7H8O2.H2O/c1-5-2-6(8)4-7(9)3-5;/h2-4,8-9H,1H3;1H2

InChI key

NBKPNAMTHBIMLA-UHFFFAOYSA-N

Other Notes

Reagent for the determination of pentose


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library