Skip to Content
MilliporeSigma

74815

Octamethyltrisiloxane

purum, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[(CH3)3SiO]2Si(CH3)2
CAS Number:
Molecular Weight:
236.53
EC Number:
203-497-4
UNSPSC Code:
12352103
PubChem Substance ID:
Beilstein/REAXYS Number:
1753063
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

>1 (vs air)

grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.384 (lit.), n20/D 1.385

bp

153 °C (lit.)

mp

−82 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

InChI

1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3

InChI key

CXQXSVUQTKDNFP-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Octamethyltrisiloxane




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library