Skip to Content
MilliporeSigma

74650

L-Norvaline

≥99.0% (NT)

Synonym(s):

(S)-(+)-2-Aminopentanoic acid, (S)-2-Aminovaleric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
EC Number:
229-543-3
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
1721162
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (NT)

optical activity

[α]20/D +25±1°, c = 10% in 20% HCl

ign. residue

≤0.05%

mp

>300 °C (lit.)

SMILES string

CCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-2-3-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

SNDPXSYFESPGGJ-BYPYZUCNSA-N

Other Notes

Competitive inhibitor of ornithine carbamyltransferase (transcarbamylase)


Still not finding the right product?

Explore all of our products under L-Norvaline




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library