Skip to Content
MilliporeSigma

60066

Potassium bis(trimethylsilyl)amide

purum, ≥95.0% (T)

Synonym(s):

Hexamethyldisilazane potassium salt, Potassium hexamethyldisilazide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[(CH3)3Si]2NK
CAS Number:
Molecular Weight:
199.48
UNSPSC Code:
12352000
Beilstein/REAXYS Number:
4006754
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥95.0% (T)

SMILES string

C[Si](C)(C)N([K])[Si](C)(C)C

InChI

1S/C6H18NSi2.K/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

IUBQJLUDMLPAGT-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library