Skip to Content
MilliporeSigma

54610

3-Hydroxybenzoic acid

puriss., ≥99.0% (T)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
EC Number:
202-726-5
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
508160
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss.

assay

≥99.0% (T)

ign. residue

≤0.07%

mp

200-203 °C (lit.), 201-205 °C

SMILES string

OC(=O)c1cccc(O)c1

InChI

1S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

IJFXRHURBJZNAO-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 3-Hydroxybenzoic acid


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library