Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
EC Number:
214-538-0
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
2054360
grade
puriss. p.a.
assay
≥98.5% (HPLC)
mp
178-181 °C (lit.)
cation traces
Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤20 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤10 mg/kg
λmax
337 nm at >2000
suitability
suitable for matrix substance for MALDI-MS
storage temp.
2-8°C
SMILES string
Oc1cccc2Cc3cccc(O)c3C(=O)c12
InChI
1S/C14H10O3/c15-10-5-1-3-8-7-9-4-2-6-11(16)13(9)14(17)12(8)10/h1-6,15-16H,7H2
InChI key
NUZWLKWWNNJHPT-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Dithranol
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.