Skip to Content
MilliporeSigma

53280

Hippuric acid

≥97.0% (T)

Synonym(s):

N-Benzoylglycine, Benzoylaminoacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CONHCH2COOH
CAS Number:
Molecular Weight:
179.17
EC Number:
207-806-3
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1073987
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97.0% (T)

mp

187-191 °C

SMILES string

OC(=O)CNC(=O)c1ccccc1

InChI

1S/C9H9NO3/c11-8(12)6-10-9(13)7-4-2-1-3-5-7/h1-5H,6H2,(H,10,13)(H,11,12)

InChI key

QIAFMBKCNZACKA-UHFFFAOYSA-N

Gene Information

rat ... Akr1b1(24192)



Still not finding the right product?

Explore all of our products under Hippuric acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library