Skip to Content
MilliporeSigma

51240

Hematoporphyrin

≥55% (HPLC)

Synonym(s):

8,13-Bis(1-hydroxyethyl)-3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H38N4O6
CAS Number:
Molecular Weight:
598.69
PubChem Substance ID:
UNSPSC Code:
12352300
EC Number:
238-450-7
MDL number:
Beilstein/REAXYS Number:
78957
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


concentration

≥55% (HPLC)

storage temp.

2-8°C

SMILES string

CC(O)c1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5CCC(O)=O)c(CCC(O)=O)c4C)c(C)c3C(C)O

Other Notes

Affinity chromatography of heme-binding proteins: Synthesis and characterization of hematin- and hematoporphyrin-agarose




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library