Skip to Content
MilliporeSigma

48975

Gliotoxin from Gliocladium fimbriatum

≥98.0% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H14N2O4S2
CAS Number:
Molecular Weight:
326.39
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
50675
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (TLC)

solubility

chloroform: 10 mg/mL, clear, colorless

antibiotic activity spectrum

viruses

mode of action

DNA synthesis | interferes, enzyme | inhibits

storage temp.

2-8°C

SMILES string

CN1C(=O)[C@]23CC4=CC=C[C@H](O)[C@H]4N2C(=O)[C@@]1(CO)SS3

InChI

1S/C13H14N2O4S2/c1-14-10(18)12-5-7-3-2-4-8(17)9(7)15(12)11(19)13(14,6-16)21-20-12/h2-4,8-9,16-17H,5-6H2,1H3/t8-,9-,12+,13+/m0/s1

InChI key

FIVPIPIDMRVLAY-RBJBARPLSA-N

General description

Chemical structure: epipolythiodioxopiperazine

Application

Noncompetitive inhibitor of the chymotrypsin-like activity of the 20S proteasome in vitro.

Other Notes

Antiphagocytic and immunomodulating agent which acts by blocking membrane thiol groups


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library