Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
FC6H4OH
CAS Number:
Molecular Weight:
112.10
EC Number:
206-681-2
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1905112
MDL number:
grade
purum
assay
≥98.0% (GC)
refractive index
n20/D 1.511 (lit.), n20/D 1.515
bp
171-172 °C/741 mmHg (lit.)
mp
16.1 °C (lit.)
density
1.256 g/mL at 25 °C (lit.)
SMILES string
Oc1ccccc1F
InChI
1S/C6H5FO/c7-5-3-1-2-4-6(5)8/h1-4,8H
InChI key
HFHFGHLXUCOHLN-UHFFFAOYSA-N
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
114.8 °F
flash_point_c
46 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

