Skip to Content
MilliporeSigma

46560

3-Fluorobenzaldehyde

purum, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
FC6H4CHO
CAS Number:
Molecular Weight:
124.11
EC Number:
207-266-9
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
970178
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (GC)

impurities

≤2% 3-fluorobenzoic acid

refractive index

n20/D 1.518

bp

66-68 °C/20 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(C=O)c1

InChI

1S/C7H5FO/c8-7-3-1-2-6(4-7)5-9/h1-5H

InChI key

PIKNVEVCWAAOMJ-UHFFFAOYSA-N



pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

132.8 °F - DIN 51755 Part 1

flash_point_c

56 °C - DIN 51755 Part 1

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library