Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(O2N)2C6H3NH2
CAS Number:
Molecular Weight:
183.12
EC Number:
210-567-8
UNSPSC Code:
12171500
MDL number:
Beilstein/REAXYS Number:
648811
grade
purum
assay
≥98.0% (HPLC)
impurities
~2% 3-nitroaniline
mp
160-162 °C
SMILES string
Nc1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C6H5N3O4/c7-4-1-5(8(10)11)3-6(2-4)9(12)13/h1-3H,7H2
InChI key
MPBZUKLDHPOCLS-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.