Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
196037
MDL number:
grade
purum
assay
≥99.0% (GC)
refractive index
n20/D 1.533
density
1.163 g/mL at 20 °C (lit.)
SMILES string
COc1c(CC=C)cc2OCOc2c1OC
InChI
1S/C12H14O4/c1-4-5-8-6-9-11(16-7-15-9)12(14-3)10(8)13-2/h4,6H,1,5,7H2,2-3H3
InChI key
LIKYNOPXHGPMIH-UHFFFAOYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Still not finding the right product?
Explore all of our products under Dillapiole
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.