Skip to Content
MilliporeSigma

37550

2,5-Dihydroxybenzoic acid

purum, ≥98.0% (T)

Synonym(s):

2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
EC Number:
207-718-5
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
2209119
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (T)

mp

203-207 °C, 204-208 °C (lit.)

SMILES string

OC(=O)c1cc(O)ccc1O

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 2,5-Dihydroxybenzoic acid




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library