Skip to Content
MilliporeSigma

37039

Fluoroimide

PESTANAL®, analytical standard

Synonym(s):

2,3-Dichloro-N-(4-fluorophenyl)maleimide, 3,4-Dichloro-1-(4-fluorophenyl)-1H-pyrrole-2,5-dione, N-(4-Fluorophenyl)dichloromaleimide, NSC 622384

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H4Cl2FNO2
CAS Number:
Molecular Weight:
260.05
PubChem Substance ID:
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1534427
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1ccc(cc1)N2C(=O)C(Cl)=C(Cl)C2=O

InChI

1S/C10H4Cl2FNO2/c11-7-8(12)10(16)14(9(7)15)6-3-1-5(13)2-4-6/h1-4H

InChI key

IPENDKRRWFURRE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library