Skip to Content
MilliporeSigma

33980

1,3-Dibromobenzene

purum, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H4Br2
CAS Number:
Molecular Weight:
235.90
EC Number:
203-574-2
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1904538
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

8.16 (vs air)

vapor pressure

5 mmHg ( 66 °C)

grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.608

bp

218-219 °C (lit.)

mp

−7 °C (lit.)

density

1.952 g/mL at 25 °C (lit.)

SMILES string

Brc1cccc(Br)c1

InChI

1S/C6H4Br2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

JSRLURSZEMLAFO-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

200.1 °F - closed cup

flash_point_c

93.4 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library