Skip to Content
MilliporeSigma

20957

Cesium bromide

puriss. p.a., ≥99.0% (AT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
232-130-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss. p.a.

assay

≥99.0% (AT)

impurities

≤0.001% total nitrogen (N)

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1000 mg/kg, sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M



Still not finding the right product?

Explore all of our products under Cesium bromide


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library