Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
EC Number:
203-305-9
UNSPSC Code:
85151701
MDL number:
Beilstein/REAXYS Number:
774687
vapor density
5.52 (vs air)
vapor pressure
1 mmHg ( 40 °C)
grade
puriss. p.a.
assay
≥99.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
refractive index
n20/D 1.413 (lit.), n20/D 1.414
bp
199 °C (lit.)
mp
−51-−50 °C (lit.)
density
1.055 g/mL at 25 °C (lit.)
application(s)
environmental
SMILES string
CCOC(=O)CC(=O)OCC
InChI
1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3
InChI key
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Diethyl malonate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.