Skip to Content
MilliporeSigma

07748

Gabapentin

pharmaceutical secondary standard; traceable to USP

Synonym(s):

1-(Aminomethyl)-cyclohexaneacetic acid, Neurontin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
EC Number:
262-076-3
UNSPSC Code:
41116000
MDL number:
Beilstein/REAXYS Number:
2359739
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

traceable to USP 1287303

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

SMILES string

NCC1(CC(O)=O)CCCCC1

InChI

1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)

InChI key

UGJMXCAKCUNAIE-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Gabapentin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library