Skip to Content
MilliporeSigma

01830

Adenine

≥99.0% (HPLC)

Synonym(s):

6-Aminopurine, Vitamin B4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
MDL number:
Beilstein/REAXYS Number:
5777
EC Number:
200-796-1
UNSPSC Code:
41106305
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (HPLC)

ign. residue

≤0.05% (as SO4)

loss

≤0.5% loss on drying, 110 °C

mp

>360 °C (lit.)

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Adenine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library