Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4280

Sigma-Aldrich

Potassium chlorate

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ first grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

5.0-6.5 (25 °C, 61.3 g/L)

mp

356 °C (lit.)

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleSkull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Ox. Sol. 1

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vicente Parra et al.
Physical chemistry chemical physics : PCCP, 9(18), 2266-2273 (2007-05-10)
Here we report on the electroactivity properties of Langmuir-Blodgett (LB) films of the hole-transport molecule 4,4',4''-tris[3-methylphenyl(phenyl)amino] triphenylamine (m-MTDATA). Fairly stable Langmuir films at the air-water interface are accomplished, despite the non-amphiphilic character of the molecule. The reflection-absorption infrared spectroscopy (RAIRS)
F Khan et al.
Biochimica et biophysica acta, 1481(2), 229-236 (2000-10-06)
Characterization of conformational transition and folding intermediates is central to the study of protein folding. We studied the effect of various alcohols (trifluoroethanol (TFE), butanol, propanol, ethanol and methanol) and salts (K(3)FeCN(6), Na(2)SO(4), KClO(4) and KCl) on the acid-induced state
Paola González Audino et al.
Pest management science, 58(2), 183-189 (2002-02-21)
The stability to heart of cis-permethrin and beta-cypermethrin in the solid phase was studied and the decomposition products identified. Samples heated at 210 degrees C in an oven in the dark showed that, in the absence of potassium chlorate (the
Sang Jin Lee et al.
Journal of biomaterials science. Polymer edition, 13(2), 197-212 (2002-05-23)
It has been recognized that adhesion and proliferation of cells on biodegradable polymers such as poly(lactic acid) (PLA), poly(glycolic acid) (PGA), and poly(lactide-co-glycolide) (PLGA) depend on the surface properties. The chloric acid (CA) treatment of these films was developed to
Tiziana Antonelli et al.
Cerebral cortex (New York, N.Y. : 1991), 15(12), 2013-2020 (2005-03-25)
The aim of this study was to investigate whether prenatal exposure to the cannabinoid CB1 receptor agonist WIN 55,212-2 (WIN) at a daily dose devoid of overt signs of toxicity and/or gross malformations (0.5 mg/kg, gestation days 5-20), influences cortical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service