Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755443

Sigma-Aldrich

N-(2-Hydroxy-3-butenyl)phthalimide

97%

Synonym(s):

2-(2-Hydroxybut-3-enyl)isoindoline-1,3-dione, 3-Hydroxy-4-phthalimido-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

75-80 °C

functional group

hydroxyl
imide

SMILES string

OC(CN1C(=O)c2ccccc2C1=O)C=C

InChI

1S/C12H11NO3/c1-2-8(14)7-13-11(15)9-5-3-4-6-10(9)12(13)16/h2-6,8,14H,1,7H2

Inchi Key

OWEYCEAOEGYVFC-UHFFFAOYSA-N

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service