Skip to Content

677531

(S,S)-DACH-pyridyl Trost ligand

97%

Synonym(s):

(+)-N,N′-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide), (1S,2S)-(+)-1,2-Bis[(2-pyridinylcarbonyl)amino]cyclohexane, (1S,2S)-1,2-Bis(2-pyridinecarboxamido)cyclohexane, (1S,2S)-N,N′-1,2-Cyclohexanediylbis(2-pyridinecarboxamide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H20N4O2
Molecular Weight:
324.38
UNSPSC Code:
12352005
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

mp

171-176 °C

SMILES string

O=C(N[C@H]1CCCC[C@@H]1NC(=O)c2ccccn2)c3ccccn3

InChI

1S/C18H20N4O2/c23-17(15-9-3-5-11-19-15)21-13-7-1-2-8-14(13)22-18(24)16-10-4-6-12-20-16/h3-6,9-14H,1-2,7-8H2,(H,21,23)(H,22,24)/t13-,14-/m0/s1

InChI key

BVYFDUZVVBXFBQ-KBPBESRZSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library