Skip to Content
MilliporeSigma

634522

Dichloroacetic acid

biotech. grade, ≥98%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Cl2CHCOOH
CAS Number:
Molecular Weight:
128.94
EC Number:
201-207-0
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1098596
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

biotech. grade

vapor density

4.5 (vs air)

vapor pressure

0.19 mmHg ( 20 °C)

assay

≥98%

form

liquid

impurities

≤0.05% water

color

APHA: <50

turbidity

0.22 NTU (clear)

refractive index

n20/D 1.466 (lit.)

bp

194 °C (lit.)

mp

9-11 °C (lit.)

density

1.563 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Dichloroacetic acid


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Oral

target_organs

Brain,Liver,Testes

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library