Skip to Content
MilliporeSigma

575062

Rhenium(III) iodide

97%

Synonym(s):

Rhenium triiodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ReI3
CAS Number:
Molecular Weight:
566.92
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
239-698-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

reaction suitability

reagent type: catalyst
core: rhenium

SMILES string

I[Re](I)I

InChI

1S/3HI.Re/h3*1H;/q;;;+3/p-3

InChI key

DDMQJDMHHOTHKW-UHFFFAOYSA-K



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library