Skip to Content
MilliporeSigma

512982

Tantalum(V) iodide

≥99.9%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
TaI5
CAS Number:
Molecular Weight:
815.47
UNSPSC Code:
12352302
EC Number:
238-742-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.9%

bp

543 °C (lit.)

mp

496 °C (lit.)

density

5.8 g/mL at 25 °C (lit.)

SMILES string

I[Ta](I)(I)(I)I



pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Inhalation - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library