Skip to Content
MilliporeSigma

465755

Silicon tetraiodide

≥99.9% trace metals basis

Synonym(s):

Tetraiodosilane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
SiI4
CAS Number:
Molecular Weight:
535.70
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
236-706-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.9% trace metals basis

reaction suitability

core: silicon

bp

287.4 °C (lit.)

mp

120.5 °C (lit.)

density

4.198 g/mL at 25 °C (lit.)

SMILES string

I[Si](I)(I)I

InChI

1S/I4Si/c1-5(2,3)4

InChI key

CFTHARXEQHJSEH-UHFFFAOYSA-N



signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library