Skip to Content
MilliporeSigma

432083

Samarium(III) fluoride

anhydrous, powder, 99.99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
SmF3
CAS Number:
Molecular Weight:
207.36
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
237-367-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

assay

99.99%

form

powder

SMILES string

F[Sm](F)F



pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library