Skip to Content
MilliporeSigma

320285

Diethyl sulfate

98%

Synonym(s):

Ethyl sulfate, Sulfuric acid diethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C2H5O)2SO2
CAS Number:
Molecular Weight:
154.18
EC Number:
200-589-6
UNSPSC Code:
12352000
PubChem Substance ID:
Beilstein/REAXYS Number:
1209714
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

5.3 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C), 2 mmHg ( 55 °C)

assay

98%

refractive index

n20/D 1.399 (lit.)

bp

208 °C (lit.)

mp

−24 °C (lit.)

density

1.177 g/mL at 25 °C (lit.)

SMILES string

CCOS(=O)(=O)OCC

InChI

1S/C4H10O4S/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3

InChI key

DENRZWYUOJLTMF-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library