268658
N,N′-Bis(salicylidene)ethylenediaminonickel(II)
98%
Synonym(s):
Nickel(II) salen, [[α,α′-(Ethylenedinitrilo)di-o-cresolato](2-)]nickel, [[2,2′-[1,2-Ethanediylbis[(nitrilo-κN)methylidyne]]bis[phenolato-κO]](2-)]nickel
About This Item
assay
98%
reaction suitability
core: nickel
reagent type: catalyst
SMILES string
C1C\N=C\c2ccccc2O[Ni]Oc3ccccc3\C=N\1
InChI
1S/C16H16N2O2.Ni/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20;/h1-8,11-12,19-20H,9-10H2;/q;+2/p-2/b17-11+,18-12+;
InChI key
DKBSGGSJQXGHBV-OYJDLGDISA-L
Application
- Covalent functionalization of multiwall carbon nanotubes
- Reduction reactions
- Epoxidation reactions
- Reduction and intramolecular cyclization
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service