Skip to Content
MilliporeSigma

128767

5-Hydroxypentanal

technical grade

Synonym(s):

5-Hydroxyvaleraldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HO(CH2)4CHO
CAS Number:
Molecular Weight:
102.13
UNSPSC Code:
12352103
EC Number:
224-165-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical grade

refractive index

n20/D 1.453 (lit.)

bp

115-122 °C/15 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

OCCCCC=O





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library