Merck
  • Mouse mammary tumor virus p75 and p110 CUX1 transgenic mice develop mammary tumors of various histologic types.
Mouse mammary tumor virus p75 and p110 CUX1 transgenic mice develop mammary tumors of various histologic types.
Cancer research (2009-09-10)
Chantal Cadieux, Valérie Kedinger, Lu Yao, Charles Vadnais, Maria Drossos, Marilène Paquet, Alain Nepveu
ABSTRACT
The p75 and p110 isoforms of the CUX1 homeodomain protein are overexpressed in breast tumors and cancer cell lines. To assess and compare the ability of these short CUX1 isoforms in driving mammary tumor development, we used site-specific transgenesis into the Hprt locus to generate transgenic mice expressing p75 or p110 CUX1 under the control of the mouse mammary tumor virus-long terminal repeat. We report that mammary tumors developed after a long latency period, and although various histopathologies were observed, the proportion of adenosquamous carcinomas was significantly higher in p75 CUX1 than in p110 CUX1 transgenic mice. Metastasis to the lung was observed in three p75 CUX1 transgenic mice. Comparisons between tumors and adjacent normal mammary glands revealed that transgenes were overexpressed in most but not all tumors, yet in all cases tested, CUX1 DNA binding was increased, suggesting that both higher expression and changes in post-translational modifications can contribute to stimulate transgene activity. Interestingly, higher expression of erbB2 mRNA was seen in most tumors, not only solid carcinomas but also adenosquamous carcinomas, whereas higher expression of various Wnt genes and activation of the beta-catenin pathway was observed primarily in adenosquamous carcinomas. Activation of erbB2 expression appeared to represent a cooperating event that occurred independently of CUX1. In contrast, chromatin immunoprecipitation, short hairpin RNA-mediated knockdown, and reporter assays established that CUX1 is involved in the transcriptional regulation of several Wnt genes. Together, these results support the notion that oncogenic activity of CUX1 can facilitate the establishment of a Wnt/beta-catenin autocrine loop.
Sigma-Aldrich
Anti-c-ErbB2/c-Neu (Ab-3) Mouse mAb (3B5), liquid, clone 3B5, Calbiochem®