- 18F-fluoromisonidazole PET/CT: a potential tool for predicting primary endocrine therapy resistance in breast cancer.
18F-fluoromisonidazole PET/CT: a potential tool for predicting primary endocrine therapy resistance in breast cancer.
Although endocrine therapy is an effective method to treat estrogen receptor (ER)-positive breast cancer, approximately 30%-40% of all hormone receptor-positive tumors display de novo resistance. The aim of our current study was to analyze whether (18)F-labeled fluoromisonidazole (1-(2-nitro-1-imidazolyl)-2-hydroxy-3-fluoropropane [(18)F-FMISO]) PET/CT could predict primary resistance to hormonal therapy in ER-positive breast cancer. Postmenopausal women who had ER-Ī±-positive breast cancer, stages II-IV, and had never received prior endocrine therapy were prospectively enrolled in this study. Patients underwent both (18)F-FDG and (18)F-FMISO PET/CT scans before and after treatment. The hottest (18)F-FDG standardized uptake value (SUV) in the tumor foci, the SUVs at 2 and 4 h, and the TBR2 h and TBR4 h for the target lesions were calculated (TBR2 h = SUV2 hT/SUV2 hB and TBR4 h = SUV4 hT/SUV4 hB [TBR is the tumor-to-background ratio]). Clinical outcomes of primary endocrine therapy with letrozole were evaluated according to the criteria of the World Health Organization after at least 3 mo of treatment. Immunohistochemistry for markers of proliferation (Ki67) and hypoxia-induced factor 1Ī± was performed on a subset of tumors that had undergone biopsy or surgery. Pearson and Spearman analysis was used to determine the correlation between the parameters of (18)F-FDG and (18)F-FMISO uptake and clinical or immunohistochemistry outcomes with a 0.01 threshold for statistical significance. A total of 45 lesions (13 primary, 32 metastatic) from 20 patients met the inclusion criteria in this study. Baseline (18)F-FDG and (18)F-FMISO PET/CT scans were obtained for 33 lesions from 16 patients. The correlation between baseline (18)F-FDG uptake and clinical outcome was weak and did not reach statistical significance (r = 0.37, P = 0.031). However, there was a significantly positive correlation between baseline (18)F-FMISO uptake (SUV2 hT, TBR2 h, SUV4 hT, and TBR4 h) and clinical outcomes after ā„3 mo of primary endocrine therapy with letrozole (r = 0.77, 0.76, 0.71, and 0.78, respectively; P < 0.0001). The application of a TBR4 h cutoff of ā„1.2 allowed the prediction of 88% of the cases of progressive disease (15/17). Despite poor correlation between (18)F-FMISO uptake and hypoxia-induced factor 1Ī± expression, a marginal positive correlation between TBR4 h and Ki67 expression was measured (r = 0.51, P = 0.011) in a subset of malignant lesions acquired by biopsy or surgery. (18)F-FMISO PET/CT can be used to predict primary endocrine resistance in ER-positive breast cancer.