Merck
Anastrozole.
Expert opinion on drug safety (2010-10-07)
Catherine M Kelly, Aman U Buzdar
ABSTRACT
Anastrozole is a third-generation aromatase inhibitor used in the adjuvant setting for the treatment of hormone receptor-positive breast cancer. Several adjuvant randomized trials have reported greater efficacy for anastrozole when compared to tamoxifen. This review discusses the mechanism of action of anastrozole; pharmacokinetic and pharmacodynamic characteristics; results of randomized controlled trials of anastrozole compared to tamoxifen in the adjuvant treatment of postmenopausal women with hormone receptor-positive breast cancer and the safety profile of anastrozole compared to tamoxifen. The reader will gain an understanding of the basic pharmacology of anastrozole, efficacy data from clinical trials comparing anastrozole to tamoxifen, safety profile of anastrozole and ongoing areas of research. The adverse effects of anastrozole.
Product Description
Anastrozole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anastrozole, ≥98% (HPLC)