Merck
  • Expression of receptor activator of NFkB (RANK) drives stemness and resistance to therapy in ER+HER2- breast cancer.
Expression of receptor activator of NFkB (RANK) drives stemness and resistance to therapy in ER+HER2- breast cancer.
Oncotarget (2020-06-02)
Inês Gomes, Bernardo P de Almeida, Sara Dâmaso, André Mansinho, Inês Correia, Sara Henriques, Raquel Cruz-Duarte, Guilherme Vilhais, Pedro Félix, Patrícia Alves, Patrícia Corredeira, Nuno L Barbosa-Morais, Luis Costa, Sandra Casimiro
ABSTRACT
The role of RANKL-RANK pathway in progesterone-driven mammary carcinogenesis and triple negative breast cancer tumorigenesis has been well characterized. However, and despite evidences of the existence of RANK-positive hormone receptor (HR)-positive breast tumors, the implication of RANK expression in HR-positive breast cancers has not been addressed before. Here, we report that RANK pathway affects the expression of cell cycle regulators and decreases sensitivity to fulvestrant of estrogen receptor (ER)-positive (ER+)/HER2- breast cancer cells, MCF-7 and T47D. Moreover, RANK overexpressing cells had a staminal and mesenchymal phenotype, with decreased proliferation rate and decreased susceptibility to chemotherapy, but were more invasive in vivo. In silico analysis of the transcriptome of human breast tumors, confirmed the association between RANK expression and stem cell and mesenchymal markers in ER+HER2- tumors. Importantly, exposure of ER+HER2- cells to continuous RANK pathway activation by exogenous RANKL, in vitro and in vivo, induced a negative feedback effect, independent of RANK levels, leading to the downregulation of HR and increased resistance to hormone therapy. These results suggest that ER+HER2- RANK-positive cells may constitute an important reservoir of slow cycling, therapy-resistance cancer cells; and that RANK pathway activation is deleterious in all ER+HER2- breast cancer cells, independently of RANK levels.
Sigma-Aldrich
Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, clone 7F9.2, from mouse
Sigma-Aldrich
Anti-Cyclin E Antibody, clone HE12, clone HE12, Upstate®, from mouse