Merck
  • Hexokinase-2 Expression in 11C-Methionine-Positive, 18F-FDG-Negative Multiple Myeloma.
Hexokinase-2 Expression in 11C-Methionine-Positive, 18F-FDG-Negative Multiple Myeloma.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2018-11-06)
Stefan Kircher, Antje Stolzenburg, Klaus Martin Kortüm, Malte Kircher, Matteo Da Via, Samuel Samnick, Andreas K Buck, Hermann Einsele, Andreas Rosenwald, Constantin Lapa
ABSTRACT
PET with 18F-FDG is the standard modality in nuclear medicine for imaging multiple myeloma (MM). However, viable MM as detected by MRI or PET with other metabolic tracers, including 11C-methionine, may be missed-for example, because of low hexokinase 2 (HK2) expression of tumor cells. The aim of this study was to further investigate potential reasons for PET false negativity. Methods: A cohort of 15 mainly pretreated patients with relapsed or refractory biopsy-proven, serologically active MM who underwent both 18F-FDG and 11C-methionine PET/CT was retrospectively analyzed. Results: In 9 of the 15 patients, 18F-FDG PET was negative in the presence of viable disease. In the remaining 6 patients, both 18F-FDG and 11C-methionine PET/CT revealed the same number of MM lesions. At immunohistochemistry, 18F-FDG-negative myeloma did not exhibit significant differences in HK2 or glucose-6-phosphatase expression from 18F-FDG-positive disease (P = 0.57 and P = 0.44, respectively). Conclusion: Beyond HK2 expression, 18F-FDG negativity in (mainly pretreated) MM patients seems to be associated with additional causes not yet known.
Sigma-Aldrich
Anti-HK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution