
Piperazine citrate

Synonym(s):
Citric acid sesquipiperazine salt hydrate, tri-Piperazine dicitrate
Linear Formula:
(C4H10N2)3 · (C6H8O7)2 · xH2O
CAS Number:
Molecular Weight:
321.33 (anhydrous basis)
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing